Skip to Content
Merck
All Photos(1)

Key Documents

V900925

Sigma-Aldrich

Glycogen from oyster

≥99%, Vetec, reagent grade, Type II

Synonym(s):

Glycogen from oyster

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

≥99%

form

powder

storage temp.

2-8°C

SMILES string

O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO[C@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)CO)O[C@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CO)O)O)O[C@@H]2[C@H](O[C@@H]([C@@H]([C@H]2O)O)O)CO

InChI

1S/C24H42O21/c25-1-5-9(28)11(30)16(35)22(41-5)39-4-8-20(45-23-17(36)12(31)10(29)6(2-26)42-23)14(33)18(37)24(43-8)44-19-7(3-27)40-21(38)15(34)13(19)32/h5-38H,1-4H2/t5-,6-,7-,8-,9-,10-,11+,12+,13-,14-,15-,16-,17-,18-,19-,20-,21+,22+,23-,24-/m1/s1

InChI key

BYSGBSNPRWKUQH-UJDJLXLFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycogen is a branched polymer of glucose synthesized by animal cells for energy storage and release. It is constructed of predominantly α1→4 glycosidic bonds with branches created through α1→6 glycosidic bonds.

Application

Oyster glycogen has been used to induce inflammation and to promote extravasation of granulocytes and neutrophils in animal models after intraperitoneal injection. Glycogen also acts as a vascular permeability probe.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service