Skip to Content
Merck
All Photos(3)

Key Documents

V900893

Sigma-Aldrich

Collagenase from Clostridium histolyticum

Type IV, Vetec, reagent grade, powder, ≥160 CDU/mg solid

Synonym(s):

Clostridiopeptidase A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

biological source

Clostridium histolyticum

grade

reagent grade

product line

Vetec

form

powder

specific activity

≥160 CDU/mg solid

technique(s)

cell culture | mammalian: suitable
single cell analysis: suitable

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Collagenase from Clostridium histolyticum is a collagenolytic enzyme. It interacts with collagen-like substances and is responsible for the degradation of collagen into small peptides.[1]

Application

Collagenase from Clostridium histolyticum may be used for the isolation of cells from many types of animal tissue. VETEC Collagenase Type IV has low tryptic activity. It is suitable for the digestion of islet cells from the pancreas.[2]

Suitable for use in preparation of single cell suspension for sequencing.

Biochem/physiol Actions

Collagenase is activated by four gram atom calcium per mole enzyme. It is inhibited by ethylene glycol-bis(beta-aminoethyl ether) - N, N, N′,N′-tetraacetic acid, beta-mercaptoethanol, glutathione, thioglycolic acid and 8-hydroxyquinoline.

Quality

Also contains clostripain, neutral protease, and tryptic activities.

Unit Definition

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C in the presence of calcium ions. One FALGPA hydrolysis unit hydrolyzes 1.0 μmole of furylacryloyl-Leu-Gly-Pro-Ala per min at 25°C. One Neutral Protease unit hydrolyzes casein to produce color equivalent to 1.0 μmole of tyrosine per 5 hr at pH 7.5 at 37°C. One Clostripain Unit hydrolyzes 1.0 μmole of BAEE per min at pH 7.6 at 25°C in the presence of DTT.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yen-Liang Li et al.
Oncoimmunology, 9(1), 1830513-1830513 (2020-10-30)
Lymph-node metastasis is a prognosis factor for poor clinical outcome of breast cancer patients. Currently, how breast cancer cells establish pre-metastatic niche in the tumor-draining lymph nodes (TDLNs) is still unclear. To address this question, we isolated heterogeneous cells including
Telomerase reverse transcriptase acts in a feedback loop with NF-KB pathway to regulate macrophage polarization in alcoholic liver disease.
Wu X Q, et al.
Scientific Reports, 6, 18685-18685 (2016)
Xiao-Qin Wu et al.
Scientific reports, 6, 18685-18685 (2016-01-05)
Activation of Kupffer cells (KCs) plays a central role in the pathogenesis of alcoholic liver disease (ALD). C57BL/6 mice fed EtOH-containing diet showed a mixed induction of hepatic classical (M1) and alternative (M2) macrophage markers. Since telomerase activation occurs at
Q Liang et al.
Clinical and experimental immunology, 203(1), 55-65 (2020-10-06)
Semi-allogeneic embryos are not rejected by the maternal immune system due to maternal-fetal immune tolerance. Progesterone (P) receptor (PR)-expressing γδ T cells are present in healthy pregnant women. In the presence of P, these cells secrete an immunomodulatory protein called
Xiancai Ma et al.
Immunity, 53(6), 1315-1330 (2020-12-05)
Various vaccine strategies have been proposed in response to the global COVID-19 pandemic, each with unique strategies for eliciting immune responses. Here, we developed nanoparticle vaccines by covalently conjugating the self-assembled 24-mer ferritin to the receptor binding domain (RBD) and/or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service