Skip to Content
Merck
All Photos(1)

Key Documents

V900735

Sigma-Aldrich

Ethyl palmitate

Vetec, reagent grade, 95%

Synonym(s):

Ethyl hexadecanoate, Palmitic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COOC2H5
CAS Number:
Molecular Weight:
284.48
Beilstein:
1782663
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
grade:
reagent grade

grade

reagent grade

product line

Vetec

Assay

95%

refractive index

n20/D 1.440 (lit.)

bp

192-193 °C/10 mmHg (lit.)

mp

24-26 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC

InChI

1S/C18H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-4-2/h3-17H2,1-2H3

InChI key

XIRNKXNNONJFQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Gabrielli et al.
Advances in colloid and interface science, 87(2-3), 75-111 (2001-02-24)
Studies performed on the interactions and reactions of compounds in the bidimensional state, essentially in monolayers and Langmuir-Blodgett multilayers, with substances in the aqueous subphase are reported. More precisely, the following is illustrated: (i) interactions between acid amphiphiles and prevalently
M A Diczfalusy et al.
European journal of biochemistry, 259(1-2), 404-411 (1999-01-23)
Fatty acid ethyl esters have been detected in high concentrations in organs commonly damaged by alcohol abuse and are regarded as being important non-oxidative metabolites of ethanol. The formation of fatty acid ethyl esters (FAEEs) has been ascribed to two
Ashanty M Pina-Rodriguez et al.
Journal of agricultural and food chemistry, 57(11), 4657-4662 (2009-05-06)
Amaranth oil is rich in linoleic, oleic, and palmitic acids. Structured lipids (SLs) with specific functional and nutritional characteristics can be prepared through chemical or enzymatic interesterification. The aim of this study was to increase the palmitic acid content at
G DePergola et al.
Alcoholism, clinical and experimental research, 15(2), 184-189 (1991-03-01)
The concentration of ethyl esters of fatty acids as well as the activity of the enzyme synthesizing these esters (fatty acid ethyl ester synthase) were determined in adipose tissue of rats ingesting ethanol (9-16 g/kg body weight/day) for different periods
Kosei Eguchi et al.
Cell metabolism, 15(4), 518-533 (2012-04-03)
Consumption of foods high in saturated fatty acids (FAs) as well as elevated levels of circulating free FAs are known to be associated with T2D. Though previous studies showed inflammation is crucially involved in the development of insulin resistance, how

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service