Skip to Content
Merck
All Photos(2)

Key Documents

V900600

Sigma-Aldrich

DL-Tartaric acid

Vetec, reagent grade, 99%

Synonym(s):

DL-2,3-Dihydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CHOH)2COOH
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725148
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

mp

210-212 °C (lit.)

SMILES string

O[C@@H]([C@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m0/s1

InChI key

FEWJPZIEWOKRBE-LWMBPPNESA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

410.0 °F - closed cup

Flash Point(C)

210 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmen-Alice Teacă et al.
Carbohydrate polymers, 93(1), 307-315 (2013-03-08)
The present paper describes the preparation and characterization of polysaccharides-based bio-composite films obtained by the incorporation of 10, 20 and 30 wt% birch cellulose (BC) within a glycerol plasticized matrix constituted by the corn starch (S) and chemical modified starch
Andrea Bencini et al.
Chemical communications (Cambridge, England), 48(84), 10428-10430 (2012-09-18)
A chiral ditopic polyammonium receptor featuring two [9]aneN(3) moieties separated by a (S)-BINOL linker is able to selectively bind and sense in water (S,S)-tartaric acid over its (R,R)/meso forms.
Anne Toboldt et al.
Applied and environmental microbiology, 78(20), 7347-7357 (2012-08-14)
In this study, the population structure, incidence, and potential sources of human infection caused by the d-tartrate-fermenting variant of Salmonella enterica serovar Paratyphi B [S. Paratyphi B (dT+)] was investigated. In Germany, the serovar is frequently isolated from broilers. Therefore
J W H Smith et al.
Journal of hazardous materials, 235-236, 279-285 (2012-09-07)
Impregnated activated carbons (IACs) that are used in multi-gas respirator applications usually contain copper and/or zinc impregnants. Co-impregnating with properly selected acids can improve the distribution of the metallic impregnant on the carbon and improve the gas adsorption capacity of
Prashant K Metri et al.
Chemistry, an Asian journal, 8(2), 488-493 (2012-12-05)
Total synthesis of the polyhydroxy caprolactam amide natural product, bengamide E, is accomplished starting from tartaric acid. Key reactions in the synthesis include desymmetrization of the bis(dimethylamide) unit of tartaric acid, Zn(BH(4))(2)-mediated anti-selective reduction, and a Horner-Wadsworth-Emmons olefination.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service