Skip to Content
Merck
All Photos(2)

Key Documents

V900500

Millipore

Agar

Vetec, reagent grade, suitable for microbiology

Synonym(s):

Agar-agar, Gum agar, Agar-agar, Gum agar

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106200

grade

reagent grade

Quality Level

sterility

non-sterile

product line

Vetec

shelf life

limited shelf life, expiry date on the label

storage condition

(Tightly closed)

pH

5-8 (1.5% in solution)

application(s)

microbiology

SMILES string

O1[C@@H]([C@@H]([C@@H]([C@H](C1OC2[C@H]3OCC2OC([C@H]3O)C)O)OC)O)CO

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Agar is widely used in microbiology as a solidifying agent in microbial culture media. It is a polysaccharide derived from seaweed and is primarily composed of agarose. Agar is a mostly inert substance, which makes it an ideal material for microbial culture media because it doesn’t interfere with the growth of microorganisms. Agar can solidify media, allowing microorganisms to grow in a solid environment, which facilitates the formation of distinct colonies and selective isolation of specific microorganisms. Agar is also used in the preparation of selective and differential media, which can provide additional nutrients or other chemical substances to promote or inhibit the growth of specific microorganisms selectively. Agar is sterilized by autoclaving and then poured into Petri dishes, where it can cool and solidify, forming the surface for microbial growth.

Application

Agar is commonly used as a solidifying agent in many microbial culture media for the growth and identification of bacteria, yeasts, and molds from various pharmaceutical, environmental, and food and beverage samples.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohua Dong et al.
Cell reports, 30(8), 2614-2626 (2020-02-27)
Synapses are fundamental to the normal function of the nervous system. Glia play a pivotal role in regulating synaptic formation. However, how presynaptic neurons assemble synaptic structure in response to the glial signals remains largely unexplored. To address this question
Amanda R Smith et al.
Journal of food protection, 76(4), 674-679 (2013-04-12)
Quality control procedures during food processing may involve direct inoculation of food samples onto appropriate selective media for subsequent enumeration. However, sublethally injured bacteria often fail to grow, enabling them to evade detection and intervention measures and ultimately threaten the
Matteo Francavilla et al.
Carbohydrate polymers, 92(2), 1555-1560 (2013-02-13)
Porous agar materials have been prepared from marine macroalgae species using a simple microwave-assisted extraction/drying methodology, providing a new family of polysaccharide derived porous solids. The microwave-assisted extraction allows a more efficient and less time-consuming extraction of the polysaccharide compared
A Ia Mukhachev et al.
Zhurnal mikrobiologii, epidemiologii, i immunobiologii, (2)(2), 28-37 (2013-06-29)
Study the influence of low temperature (cold) electrolyte plasma (CEP) on survivability of some mycoplasma strains growing in agar as well as mycoplasma that most frequently contaminate transplantable human cell lines of normal and malignant origin with the aim of
Xiaomin Liu et al.
Medical image computing and computer-assisted intervention : MICCAI ... International Conference on Medical Image Computing and Computer-Assisted Intervention, 15(Pt 1), 373-380 (2013-01-05)
Automatically detecting and tracking the motion of Myxococcus xanthus bacteria provide essential information for studying bacterial cell motility mechanisms and collective behaviors. However, this problem is difficult due to the low contrast of microscopy images, cell clustering and colliding behaviors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service