Skip to Content
Merck
All Photos(3)

Key Documents

V900497

Sigma-Aldrich

Pepsin from porcine gastric mucosa

Vetec, reagent grade

Synonym(s):

Pepsin A, Pepsin from hog stomach

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

solid

specific activity

400-800 units/mg protein

mol wt

35 kDa

Looking for similar products? Visit Product Comparison Guide

Application

Pepsin cleavage can be used to produce F(ab′)2 fragments of antibodies. pepsin at www.sigma-aldrich.com/enzymeexplorer.

Biochem/physiol Actions

Preferential cleavage: hydrophobic and aromatic residues in P1 and P1′ postitions. Cleaves Phe-Val, Gln-His, Glu-Ala, Ala-Leu, Leu-Tyr, Tyr-Leu, Gly-Phe, Phe-Phe and Phe-Tyr bonds in the β chain of insulin

Unit Definition

One unit will produce a change in A280 of 0.001 per minute at pH 2.0 at 37 deg C, measured as TCA-Soluble products using hemoglobin as substrate.

Analysis Note

Optimum pH is 2-4. Active in 4 M urea and 3 M guanidine HCl. Stable at 60 °C. Pepsin is irreversibly inactivated at pH 8.0 - 8.5.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liu Lin et al.
Frontiers in nutrition, 9, 833712-833712 (2022-03-01)
The effects of condiment (salt) and processing technic (homogenization) on digestion and interfacial properties of micro/nano-sized colloidal particles (MNCPs) in bigeye tuna head soup (BTHS) using simulated gastrointestinal digestion model in vitro were investigated. For MNCPs in BTHS, the triglycerides
Chuan-Hsiao Han et al.
Food chemistry, 138(2-3), 923-930 (2013-02-16)
Our previous report showed that yam dioscorin and its peptic hydrolysates exhibit radical scavenging activities; however, the functions of these peptic hydrolases are still under investigation. In this study, the thiol-containing peptides derived from computer-aided simulation of pepsin hydrolysis of
Mahmoud S Ali et al.
The Laryngoscope, 123(2), 434-439 (2012-10-17)
To assess if, as previously reported in the literature, bile acids inhibit pepsin activity, resulting in pepsin having a less important role in laryngopharyngeal damage in reflux disease. Prospective translational research study. A total of 78 patient's fasting gastric juice
María Llarena-Reino et al.
Veterinary parasitology, 191(3-4), 276-283 (2012-10-10)
During the last 50 years human anisakiasis has been rising while parasites have increased their prevalence at determined fisheries becoming an emergent major public health problem. Although artificial enzymatic digestion procedure by CODEX (STAN 244-2004: standard for salted Atlantic herring
Adrian Allen et al.
American journal of physiology. Cell physiology, 288(1), C1-19 (2004-12-14)
Secretion of bicarbonate into the adherent layer of mucus gel creates a pH gradient with a near-neutral pH at the epithelial surfaces in stomach and duodenum, providing the first line of mucosal protection against luminal acid. The continuous adherent mucus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service