Skip to Content
Merck
All Photos(3)

Key Documents

V900480

Sigma-Aldrich

CHAPS

Vetec, reagent grade, 96%

Synonym(s):

3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H58N2O7S
CAS Number:
Molecular Weight:
614.88
MDL number:
UNSPSC Code:
12161703
PubChem Substance ID:

grade

reagent grade

description

zwitterionic

product line

Vetec

Assay

96%

mol wt

micellar avg mol wt 6150

aggregation number

10

CMC

6 mM (20-25°C)

transition temp

cloud point >100 °C

SMILES string

C[C@H](CCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)CC4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C32H58N2O7S/c1-21(8-11-29(38)33-14-6-15-34(4,5)16-7-17-42(39,40)41)24-9-10-25-30-26(20-28(37)32(24,25)3)31(2)13-12-23(35)18-22(31)19-27(30)36/h21-28,30,35-37H,6-20H2,1-5H3,(H-,33,38,39,40,41)/t21-,22?,23-,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

UMCMPZBLKLEWAF-RFCNGIAKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

CHAPS is a nondenaturing zwitterionic detergent for membrane biochemistry.
Useful for solubilizing membrane proteins and breaking protein-protein interactions. CHAPS′ small micellar molecular weight (6,150) and high critical micelle concentration (6-10 mM) allow it to be removed from samples by dialysis. It is also suitable for protein solubilization for isoelectric focusing and two-dimensional electrophoresis. CHAPS is commonly used for non-denaturing (without urea) IEF and has been shown to give excellent resolution of some subcellular preparations and plant proteins. Concentrations between 2-4% (w/v) are typically used in an IEF gel.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hannah R Bridges et al.
The Biochemical journal, 462(3), 475-487 (2014-07-16)
The biguanide metformin is widely prescribed for Type II diabetes and has anti-neoplastic activity in laboratory models. Despite evidence that inhibition of mitochondrial respiratory complex I by metformin is the primary cause of its cell-lineage-specific actions and therapeutic effects, the molecular
Hendrik Wesseling et al.
Journal of proteome research, 14(1), 411-421 (2014-11-05)
There is substantial interest in the N-methyl-d-aspartate (NMDA) receptor antagonist ketamine in psychiatric research because it exerts acute psychotomimetic and rapid antidepressant effects in rodents and humans. Here, we investigated proteomic changes in brain and serum after acute treatment of
Bernardo S Franklin et al.
Nature immunology, 15(8), 727-737 (2014-06-24)
Microbes or danger signals trigger inflammasome sensors, which induce polymerization of the adaptor ASC and the assembly of ASC specks. ASC specks recruit and activate caspase-1, which induces maturation of the cytokine interleukin 1β (IL-1β) and pyroptotic cell death. Here
Stephanie Campbell et al.
Journal of virology, 88(15), 8667-8677 (2014-05-23)
Apoptosis is a tightly regulated process that plays a crucial role in the removal of virus-infected cells, a process controlled by both pro- and antiapoptotic members of the Bcl-2 family. The proapoptotic proteins Bak and Bax are regulated by antiapoptotic
Laetitia Aerts et al.
The Journal of general virology, 96(Pt 4), 767-774 (2014-12-19)
The human metapneumovirus (HMPV) fusion (F) protein is the most immunodominant protein, yet subunit vaccines containing only this protein do not confer complete protection. The HMPV matrix (M) protein induces the maturation of antigen-presenting cells in vitro. The inclusion of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service