Skip to Content
Merck
All Photos(3)

Key Documents

V900476

Sigma-Aldrich

TES

Vetec, reagent grade, 99%

Synonym(s):

2-[(2-Hydroxy-1,1-bis(hydroxymethyl)ethyl)amino]ethanesulfonic acid, N-[Tris(hydroxymethyl)methyl]-2-aminoethanesulfonic acid, TES free acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15NO6S
CAS Number:
Molecular Weight:
229.25
Beilstein:
1957061
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

99%

form

crystalline powder

useful pH range

6.8-8.2

pKa (25 °C)

7.5

mp

~225 °C (dec.)

solubility

water: 0.5 g/mL, clear, colorless

SMILES string

OCC(CO)(CO)NCCS(O)(=O)=O

InChI

1S/C6H15NO6S/c8-3-6(4-9,5-10)7-1-2-14(11,12)13/h7-10H,1-5H2,(H,11,12,13)

InChI key

JOCBASBOOFNAJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R H Martin et al.
Biology of reproduction, 47(2), 268-270 (1992-08-01)
Human sperm karyotypes can be prepared after fusion of human sperm with Golden hamster oocytes. Most laboratories use one of two methods of sperm capacitation: incubation of freshly-ejaculated sperm in Biggers, Whitten, and Whittingham (BWW) medium for 5-7 h at
Yikang Wu et al.
Organic letters, 4(13), 2141-2144 (2002-06-21)
[reaction: see text] In DMSO cleavage of triethylsilyl (TES) ethers by o-iodoxybenzoic acid (IBX) was significantly faster than cleavage of tert-butyldimethylsilyl (TBS) ethers or further oxidation into carbonyl compounds. In most cases, TES protecting groups could be removed in good
M J Taylor et al.
The British journal of ophthalmology, 73(10), 781-791 (1989-10-01)
Preservation solutions for short-term storage of isolated donor corneas for use in penetrating keratoplasty have all been based on tissue culture medium, on the assumption that media designed to maintain the viability of cells at physiological temperatures will also provide
N Sugiyama et al.
Molecular reproduction and development, 58(1), 30-38 (2001-01-06)
The growth, differentiation, and death/survival of spermatogonia are precisely regulated for the proper production of spermatozoa. We have previously shown that Bcl-2 ectopically expressed in spermatogonia caused the inhibition of normal spermatogonial apoptosis and the subsequent failure of differentiation in
Marcel A L M van Assen et al.
Psychological methods, 20(3), 293-309 (2014-11-18)
Publication bias threatens the validity of meta-analytic results and leads to overestimation of the effect size in traditional meta-analysis. This particularly applies to meta-analyses that feature small studies, which are ubiquitous in psychology. Here we develop a new method for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service