Skip to Content
Merck
All Photos(2)

Documents

V900464

Sigma-Aldrich

Sodium butyrate

Vetec, reagent grade, 99%

Synonym(s):

Butyric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COONa
CAS Number:
Molecular Weight:
110.09
Beilstein:
3629439
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

form

powder

mp

250-253 °C (lit.)

solubility

water: 100 mg/mL, clear to slightly hazy, colorless

SMILES string

[Na+].CCCC([O-])=O

InChI

1S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1

InChI key

MFBOGIVSZKQAPD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Autolyzed yeast and sodium butyrate supplemented alone to diets promoted improvements in performance, intestinal health and nutrient transporter in weaned piglets.: This study found that the supplementation of autolyzed yeast and sodium butyrate in the diet of weaned piglets improved their performance, intestinal health, and nutrient absorption. These findings suggest potential applications in enhancing animal health and productivity in agricultural settings (Correia et al., 2024).

  • The New Buffer Salt-Protected Sodium Butyrate Promotes Growth Performance by Improving Intestinal Histomorphology, Barrier Function, Antioxidative Capacity, and Microbiota Community of Broilers.: This study demonstrated that a new formulation of buffer salt-protected sodium butyrate significantly improved growth performance, intestinal health, and microbiota balance in broilers. Such advancements could enhance poultry production efficiency and health management (Melaku et al., 2024).

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ras Trokovic et al.
Stem cell research, 15(1), 254-262 (2015-06-23)
Somatic cells can be reprogrammed into induced pluripotent stem cells (iPSC) by the forced expression of the transcription factors OCT4, SOX2, KLF4 and c-MYC. Pluripotent reprogramming appears as a slow and inefficient process because of genetic and epigenetic barriers of
Regina Kanski et al.
Journal of cell science, 127(Pt 20), 4368-4380 (2014-08-17)
Glial fibrillary acidic protein (GFAP) is the main intermediate filament in astrocytes and is regulated by epigenetic mechanisms during development. We demonstrate that histone acetylation also controls GFAP expression in mature astrocytes. Inhibition of histone deacetylases (HDACs) with trichostatin A
Duy Anh Bui et al.
Journal of chromatography. A, 1421, 203-208 (2015-06-21)
A new absorbance detector for capillary electrophoresis featuring relatively high intensity light-emitting diodes as radiation sources and photodiodes for the deep-UV range was developed. The direct relationship of absorbance values and concentrations was obtained by emulating Lambert-Beer's law with the
Katrin Töpfer et al.
Journal of immunology (Baltimore, Md. : 1950), 194(7), 3201-3212 (2015-03-06)
NK cells are emerging as new effectors for immunotherapy of cancer. In particular, the genetic engraftment of chimeric Ag receptors (CARs) in NK cells is a promising strategy to redirect NK cells to otherwise NK cell-resistant tumor cells. On the
Nina A Sibbesen et al.
Oncotarget, 6(24), 20555-20569 (2015-08-06)
Aberrant activation of Janus kinase-3 (Jak3) and its key down-stream effectors, Signal Transducer and Activator of Transcription-3 (STAT3) and STAT5, is a key feature of malignant transformation in cutaneous T-cell lymphoma (CTCL). However, it remains only partially understood how Jak3/STAT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service