Skip to Content
Merck
All Photos(2)

Key Documents

V900462

Sigma-Aldrich

Cytosine

Vetec, reagent grade, 99%

Synonym(s):

4-Amino-2-hydroxypyrimidine, 4-Aminopyrimidin-2-(1H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N3O
CAS Number:
Molecular Weight:
111.10
Beilstein:
2637
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

form

powder

mp

>300 °C (lit.)

solubility

0.5 M HCl: 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

SMILES string

NC1=NC(=O)NC=C1

InChI

1S/C4H5N3O/c5-3-1-2-6-4(8)7-3/h1-2H,(H3,5,6,7,8)

InChI key

OPTASPLRGRRNAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irene M Kaplow et al.
Genome research, 25(6), 907-917 (2015-04-26)
DNA methylation is an epigenetic modification that plays a key role in gene regulation. Previous studies have investigated its genetic basis by mapping genetic variants that are associated with DNA methylation at specific sites, but these have been limited to
Ryan Lister et al.
Science (New York, N.Y.), 341(6146), 1237905-1237905 (2013-07-06)
DNA methylation is implicated in mammalian brain development and plasticity underlying learning and memory. We report the genome-wide composition, patterning, cell specificity, and dynamics of DNA methylation at single-base resolution in human and mouse frontal cortex throughout their lifespan. Widespread
Kian Peng Koh et al.
Current opinion in cell biology, 25(2), 152-161 (2013-03-19)
Changes in cellular phenotypes and identities are fundamentally regulated by epigenetic mechanisms including DNA methylation, post-translational histone modifications and chromatin remodeling. Recent genome-wide profiles of the mammalian DNA 'methylome' suggest that hotspots of dynamic DNA methylation changes during cell fate
Rahul M Kohli et al.
Nature, 502(7472), 472-479 (2013-10-25)
DNA methylation has a profound impact on genome stability, transcription and development. Although enzymes that catalyse DNA methylation have been well characterized, those that are involved in methyl group removal have remained elusive, until recently. The transformative discovery that ten-eleven
Gitali Devi et al.
Nucleic acids research, 42(6), 4008-4018 (2014-01-16)
Peptide nucleic acids (PNAs) have been developed for applications in biotechnology and therapeutics. There is great potential in the development of chemically modified PNAs or other triplex-forming ligands that selectively bind to RNA duplexes, but not single-stranded regions, at near-physiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service