Skip to Content
Merck
All Photos(3)

Key Documents

V900408

Sigma-Aldrich

L-Glutamic acid

≥99%, Vetec

Synonym(s):

(S)-2-Aminopentanedioic acid, Glu

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
147.13
Beilstein:
1723801
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Pricing and availability is not currently available.

Product Name

L-Glutamic acid, Vetec, reagent grade, ≥99%

grade

reagent grade

product line

Vetec

Assay

≥99%

form

powder

color

white to off-white

mp

205 °C (dec.) (lit.)

density

1.54 g/cm3 at 20 °C

SMILES string

N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1

InChI key

WHUUTDBJXJRKMK-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

An excitatory amino acid neurotransmitter that is an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA).

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luigi F Agnati et al.
Pharmacological reviews, 55(3), 509-550 (2003-07-19)
The molecular basis for the known intramembrane receptor/receptor interactions among G protein-coupled receptors was postulated to be heteromerization based on receptor subtype-specific interactions between different types of receptor homomers. The discovery of GABAB heterodimers started this field rapidly followed by
G Alfredsson et al.
Biological psychiatry, 23(7), 689-697 (1988-04-01)
The concentrations of homovanillic acid (HVA), 5-hydroxyindoleacetic acid (5-HIAA), 4-hydroxy-3-methoxyphenylglycol (HMPG), and glutamate were determined in cerebrospinal fluid (CSF) and serum in 10 healthy volunteers. The monoamine metabolites were measured by mass fragmentography and the glutamate by high-performance liquid chromatography.
Katarzyna Rogoz et al.
Molecular pharmacology, 85(2), 322-334 (2013-11-28)
The transient receptor potential cation channel subfamily V member 1 (TRPV1) is known as a thermosensor and integrator of inflammation-induced hyperalgesia. TRPV1 is expressed in a subpopulation of primary afferent neurons that express several different neurotransmitters. The role of the
Kimberly J Dougherty et al.
Neuron, 80(4), 920-933 (2013-11-26)
Locomotion is controlled by spinal networks that generate rhythm and coordinate left-right and flexor-extensor patterning. Defined populations of spinal interneurons have been linked to patterning circuits; however, neurons comprising the rhythm-generating kernel have remained elusive. Here, we identify an ipsilaterally
Nils Muhlert et al.
Journal of neurology, neurosurgery, and psychiatry, 85(8), 833-839 (2014-01-17)
Glutamate is the principal excitatory neurotransmitter and is involved in normal brain function. Cognitive impairment is common in multiple sclerosis (MS), and understanding its mechanisms is crucial for developing effective treatments. We used structural and metabolic brain imaging to test

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service