Skip to Content
Merck

V900400

L-Cysteine

Vetec, reagent grade, 97%

Synonym(s):

(R)-2-Amino-3-mercaptopropionic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HSCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
121.16
EC Number:
200-158-2
UNSPSC Code:
12352209
PubChem Substance ID:
Beilstein/REAXYS Number:
1721408
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

reagent grade

product line

Vetec

assay

97%

form

crystalline powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

240 °C (dec.) (lit.)

SMILES string

N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/t2-/m0/s1

InChI key

XUJNEKJLAYXESH-REOHCLBHSA-N

Biochem/physiol Actions

NMDA glutamatergic receptor agonist.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under L-Cysteine


Storage Class

13 - Non Combustible Solids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library