Skip to Content
Merck
All Photos(1)

Key Documents

V900337

Sigma-Aldrich

Iodonitrotetrazolium chloride

Vetec, reagent grade, 95%

Synonym(s):

2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, p-Iodonitrotetrazolium Violet, INT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13ClIN5O2
CAS Number:
Molecular Weight:
505.70
Beilstein:
4093224
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

95%

mp

240 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Cl-].[O-][N+](=O)c1ccc(cc1)-[n+]2nc(nn2-c3ccc(I)cc3)-c4ccccc4

InChI

1S/C19H13IN5O2.ClH/c20-15-6-8-16(9-7-15)23-21-19(14-4-2-1-3-5-14)22-24(23)17-10-12-18(13-11-17)25(26)27;/h1-13H;1H/q+1;/p-1

InChI key

JORABGDXCIBAFL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Electron acceptor for the colorimetric assay of various dehydrogenases

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - STOT SE 2 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Mermillod-Blondin et al.
Water research, 39(9), 1687-1698 (2005-05-19)
Biogeochemical processes mediated by microorganisms in river sediments (hyporheic sediments) play a key role in river metabolism. Because biogeochemical reactions in the hyporheic zone are often limited to the top few decimetres of sediments below the water-sediment interface, slow filtration
T J Mankelow et al.
The Biochemical journal, 358(Pt 2), 315-324 (2001-08-22)
Diethyl pyrocarbonate (DEPC), a histidine-modifying reagent, has been utilized to demonstrate the importance of histidine residues in the functioning of proteins. In previous studies of the NADPH oxidase, histidine residues have been determined to be important in the ability of
Mwinyikione Mwinyihija et al.
Archives of environmental contamination and toxicology, 50(3), 316-324 (2006-01-05)
A study was conducted to investigate the sediment health and water quality of the River Sagana, Kenya, as impacted by the local tanning industry. Chemical analysis identified the main chemical pollutants (pentachlorophenols and chromium) while a bioassay addressed pollutant bioavailability.
Yang Hongwei et al.
Ecotoxicology and environmental safety, 59(2), 249-255 (2004-08-26)
Anaerobic biodegradability of organic compounds can be assessed from the changes in organic compounds, the end products, and the activity of microorganisms. In this study, all three of these variables were considered to integrate assessment of the anaerobic biodegradability of
Xiao-mei Xie et al.
Journal of environmental sciences (China), 16(2), 252-255 (2004-05-13)
The impact of triazophos, bensulfuron-methyl, chlobenthiazone on soil biochemical characteristics in a paddy soil under controlled moisture (flooded soil) and temperature (25 degrees C) condition was studied. The electron transport system (ETS)/dehydrogenase activity displayed a negative correlation with triazophos, bensulfuron-methyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service