Skip to Content
Merck
All Photos(3)

Key Documents

V900304

Sigma-Aldrich

Dodecyltrimethylammonium bromide

Vetec, reagent grade, 98%

Synonym(s):

Lauryltrimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11N(CH3)3Br
CAS Number:
Molecular Weight:
308.34
Beilstein:
3597463
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

mol wt

308.34 g/mol

mp

246 °C (dec.) (lit.)

application(s)

microbiology

SMILES string

[Br-].CCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C15H34N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1H/q+1;/p-1

InChI key

XJWSAJYUBXQQDR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Long-Term Thermal Stability of Ionic Surfactants for Improving Oil Production at High-Salinity High-Temperature Conditions.: This study evaluates the effectiveness of Dodecyltrimethylammonium bromide and similar surfactants under extreme conditions, aiming to enhance oil recovery processes, demonstrating the surfactant′s robustness and applicability in the petroleum industry (Hou et al., 2024).

  • Kinetic and adsorption isotherm studies of Malachite Green dye onto surfactant-tailored alginate hydrogel beads: An influence of surfactant hydrophobicity.: This research explores the interaction between hydrophobic surfactants like Dodecyltrimethylammonium bromide and dye molecules, offering insights into environmental cleanup techniques through enhanced adsorption processes (Malik et al., 2024).

  • Role of Alkyl Chain Length in Surfactant-Induced Precipitation of Reactive Brilliant Blue KN-R.: Investigates how variations in surfactant chain length affect dye precipitation, with Dodecyltrimethylammonium bromide serving as a key component in understanding surfactant behavior in industrial dyeing processes (Liu et al., 2024).

  • Additive-anchored thermoresponsive nanoscale self-assembly generation in normal and reverse Tetronics.: Examines the role of additives like Dodecyltrimethylammonium bromide in creating responsive self-assembling materials, which are critical for developing smart materials and nanotechnology applications (Patel et al., 2024).

  • Observation of Electrostatically Driven Surface Adsorption in Mixed Surfactant Systems.: This paper details the effects of mixed surfactant systems, including Dodecyltrimethylammonium bromide, on surface adsorption phenomena, crucial for advancements in coatings and surface treatment technologies (Vilangottunjalil et al., 2024).


Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sriram Sokalingam et al.
PloS one, 7(7), e40410-e40410 (2012-07-14)
Two positively charged basic amino acids, arginine and lysine, are mostly exposed to protein surface, and play important roles in protein stability by forming electrostatic interactions. In particular, the guanidinium group of arginine allows interactions in three possible directions, which
A A Mikhailovskaya et al.
The journal of physical chemistry. B, 115(33), 9971-9979 (2011-07-26)
The dynamic dilatational surface elasticity of mixed solutions of globular proteins (β-lactoglobulin (BLG) and bovine serum albumin (BSA)) with cationic (dodecyltrimethylammonium bromide (DTAB)) and anionic (sodium dodecyl sulfate (SDS)) surfactants was measured as a function of the surfactant concentration and
Adhimoorthy Prasannan et al.
Langmuir : the ACS journal of surfaces and colloids, 27(2), 766-773 (2010-12-16)
A novel synthesis of "hairy urchin"-shaped polyaniline (PAni) and its surface coverage with nanospikes was achieved from a simple microemulsion polymerization technique in the presence of β-cyclodextrin (β-CD). The rodlike micelle phase was characterized, and the key factors affecting the
I Echavarri Franco et al.
Langmuir : the ACS journal of surfaces and colloids, 28(10), 4815-4828 (2012-02-16)
The binding of cationic surfactants with varying alkyl chain length to a regiorandom conjugated polyanion, poly(3-thiophene acetic acid) (PTAA), is studied in an aqueous buffer by using absorption and emission spectroscopies, photon correlation spectroscopy, isothermal titration calorimetry, and cryogenic transmission
Marta Baptista et al.
Fertility and sterility, 99(3), 705-712 (2012-12-12)
To determine the mechanisms of spermicidal activity by cationic compounds. In vitro study with human sperm. Academic research institute in collaboration with a university hospital. Normozoospermic men providing samples for routine analysis or assisted reproductive technologies. In vitro incubation of sperm with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service