Skip to Content
Merck
All Photos(3)

Key Documents

V900269

Sigma-Aldrich

Calcium chloride dihydrate

Vetec, reagent grade, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
reagent grade
form:
powder or chunks
Pricing and availability is not currently available.

grade

reagent grade

vapor pressure

0.01 mmHg ( 20 °C)

product line

Vetec

Assay

99%

form

powder or chunks

pH

5-8 (25 °C, 147 g/L)

mp

176 °C (dec.) (lit.)

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Geetha et al.
Ecotoxicology and environmental safety, 122, 17-23 (2015-07-15)
Population explosion, depletion of water resources and prolonged droughts and floods due to climatic change lead to scarcity of pure and hygienic drinking water in most of the developing countries. Recently nanomaterials attained considerable attention as biosorbent for water purification
Avinash A Kadam et al.
Bioresource technology, 132, 276-284 (2013-02-16)
Pretreatments to sugarcane bagasse (SCB) such as CaCl2, alkali, ammonia, steam and milling showed 91%, 46%, 47%, 42% and 56% adsorption of Solvent Red 5B (SR5B); 92%, 57%, 58%, 56% and 68% adsorption of simulated dyes mixture (SDM), and 86%
Fei Ma et al.
Meat science, 95(1), 22-26 (2013-05-07)
The effects of high pressure processing (HPP) on the water-binding capacity and texture profile (TPA) of salt-soluble meat protein (SSMP) containing 0.2% CaCl2 and 0.6% κ-carrageenan (SSMP-CK) gels were investigated. The results showed that 300-400 MPa improved water-binding capacity and
Hirotada Hirama et al.
Langmuir : the ACS journal of surfaces and colloids, 29(2), 519-524 (2012-12-14)
We report a simple method for forming monodispersed, uniformly shaped gel microbeads with precisely controlled sizes. The basis of our method is the placement of monodispersed sodium alginate droplets, formed by a microfluidic device, on an agarose slab gel containing
Ioannis Parastatidis et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(10), 2389-2396 (2013-08-21)
Elevated levels of oxidative stress have been reported in abdominal aortic aneurysms (AAA), but which reactive oxygen species promotes the development of AAA remains unclear. Here, we investigate the effect of hydrogen peroxide (H2O2)-degrading enzyme catalase on the formation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service