Skip to Content
Merck
All Photos(1)

Key Documents

V900238

Sigma-Aldrich

Diethylene glycol monoethyl ether

Vetec, reagent grade, 99%

Synonym(s):

2-(2-Ethoxyethoxy)ethanol, CARBITOL, Diethylene glycol ethyl ether, Ethyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
134.17
Beilstein:
1736441
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

grade

reagent grade

vapor density

4.63 (vs air)

vapor pressure

0.12 mmHg ( 20 °C)

product line

Vetec

Assay

99%

form

liquid

expl. lim.

1.2 %, 135 °F
23.5 %, 182 °F

impurities

≤0.10% water
water

refractive index

n20/D 1.427 (lit.)

bp

202 °C (lit.)

mp

-80 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

SMILES string

CCOCCOCCO

InChI

1S/C6H14O3/c1-2-8-5-6-9-4-3-7/h7H,2-6H2,1H3

InChI key

XXJWXESWEXIICW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CARBITOL is a trademark of The Dow Chemical Company or an affiliated company of Dow
Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhupinder Singh et al.
Drug delivery, 18(8), 599-612 (2011-10-20)
Carvedilol, a widely prescribed cardiovascular drug for hypertension and congestive heart failure, exhibits low and variable bioavailability owing to poor absorption and extensive hepatic first-pass metabolism. The current research work, therefore, entails formulation development of liquid self-nano-emulsifying drug delivery systems
Rania M Hathout et al.
Molecular pharmaceutics, 7(4), 1266-1273 (2010-06-16)
This research determined the uptake of individual components of topically applied microemulsions into the stratum corneum (SC) and assessed their molecular effects on skin barrier function. The microemulsions comprised oleic acid, Tween20, Transcutol and water. The effects of selected formulations
J-L Delongeas et al.
Regulatory toxicology and pharmacology : RTP, 57(2-3), 284-290 (2010-03-30)
Drug safety research is frequently faced with the challenge of the selection of appropriate vehicles for use in in vivo non-clinical safety assessment studies. Reported here are the results of blend Labrasol, Labrafil and Transcutol, [L/L/T, (4/4/2, v/v/v)], excipients used
Maria Manconi et al.
International journal of pharmaceutics, 412(1-2), 37-46 (2011-05-03)
The ability of a recently developed novel class of liposomes to promote dermal delivery of tretinoin (TRA) was evaluated. New penetration enhancer-containing vesicles (PEVs) were prepared adding to conventional phosphatidylcholine vesicles (control liposomes) different hydrophilic penetration enhancers: Oramix NS10 (OrNS10)
Jeoung Hee Yoo et al.
Archives of pharmacal research, 33(3), 417-426 (2010-04-03)
Self-nanoemulsifying drug delivery system (SNEDDS) containing oil (Phosal 53 MCT), surfactant (Labrasol), and cosurfactant (Transcutol-HP or Lutrol-E400) was prepared to enhance solubility and dissolution of lutein. Ternary phase diagram of the SNEDDS was constructed to identify the self-emulsifying regions following

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service