Skip to Content
Merck
All Photos(1)

Key Documents

V900185

Sigma-Aldrich

Acetylsalicylic acid

Vetec, reagent grade, ≥99%

Synonym(s):

2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CH3CO2)C6H4CO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
779271
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥99%

form

powder

mp

134-136 °C (lit.)

solubility

ethanol: 50 mg/mL, clear, colorless

SMILES string

CC(=O)Oc1ccccc1C(O)=O

InChI

1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)

InChI key

BSYNRYMUTXBXSQ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Blocks the production of prostaglandins by inhibiting cyclooxygenase (prostaglandin H synthase), with greater selectivity toward the COX-1 isoform. The antithrombotic effect is due to the inhibition of COX-1 in platelets that blocks thromboxane production and platelet aggregation. Chemopreventive against colorectal and other solid tumors.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

482.0 °F - closed cup

Flash Point(C)

250 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrizio D'Ascenzo et al.
The American journal of cardiology, 115(9), 1185-1193 (2015-03-24)
The optimal antiaggregant therapy after coronary stenting in patients receiving oral anticoagulants (OACs) is currently debated. MEDLINE and Cochrane Library were searched for studies reporting outcomes of patients who underwent PCI and who were on triple therapy (TT) or dual-antiplatelet
Connie N Hess et al.
Journal of the American College of Cardiology, 66(7), 777-787 (2015-08-14)
Bleeding limits anticoagulant treatment in patients with acute coronary syndromes (ACS). We investigated whether background concomitant antiplatelet therapy influences the effects of apixaban after ACS. This study examined high-risk ACS patients who were treated with aspirin or aspirin plus clopidogrel
Christina Perneby et al.
Thrombosis and haemostasis, 95(4), 652-658 (2006-04-08)
Aspirin is widely used, but dosages in different clinical situations and the possible importance of "aspirin resistance" are debated. We performed an open cross-over study comparing no treatment (baseline) with three aspirin dosage regimens--37.5 mg/day for 10 days, 320 mg/day
Andrew O Maree et al.
Journal of the American College of Cardiology, 46(7), 1258-1263 (2005-10-04)
We investigated whether use of low-dose enteric-coated (EC) aspirin for secondary prevention of cardiovascular events has sufficient bioavailability to achieve complete platelet cyclooxygenase (COX) inhibition in all individuals. Aspirin reduces cardiovascular morbidity and mortality in patients with pre-existing vascular disease;
A O Maree et al.
Journal of thrombosis and haemostasis : JTH, 3(10), 2340-2345 (2005-09-10)
Aspirin (acetylsalicylic acid) irreversibly inhibits platelet cyclooxygenase (COX)-1, the enzyme that converts arachidonic acid (AA) to the potent platelet agonist thromboxane (TX) A2. Despite clear benefit from aspirin in patients with cardiovascular disease (CAD), evidence of heterogeneity in the way

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service