Skip to Content
Merck
All Photos(1)

Key Documents

V900088

Sigma-Aldrich

1-Methylimidazole

Vetec, reagent grade, 98%

Synonym(s):

N-Methylimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
Beilstein:
105197
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor pressure

0.4 mmHg ( 20 °C)

product line

Vetec

Assay

98%

autoignition temp.

977 °F

expl. lim.

15.7 %

refractive index

n20/D 1.495 (lit.)

bp

198 °C (lit.)

mp

−6 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

Cn1ccnc1

InChI

1S/C4H6N2/c1-6-3-2-5-4-6/h2-4H,1H3

InChI key

MCTWTZJPVLRJOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengfei Zhang et al.
Journal of chromatography. A, 1218(22), 3459-3465 (2011-04-19)
The development of mixed-mode stationary phase to achieve multiple separation capabilities in one column is very important for high performance liquid chromatography. In this paper, a new specific stationary phase based on grafting N-methylimidazolium to a monolithic silica column was
Zhan-Yong Wang et al.
The Journal of organic chemistry, 77(15), 6608-6614 (2012-07-19)
A well-defined NHC-Pd(II)-Im complex 1 was found to be an effective catalyst for the Suzuki-Miyaura coupling of aryl sulfonates including tosylates and phenylsulfonates with arylboronic acids, giving the desired coupling products in good to high yields. Acceptable yields can also
Bhaskar Sharma et al.
The journal of physical chemistry. A, 115(10), 1971-1984 (2011-02-22)
Quantum chemical [MP2(FULL)/6-311++G-(d,p)] calculations are done on the binding of hydrated Li(+), Na(+), K(+), Mg(2+), Cu(+), and Zn(2+) metal ions with biologically relevant heteroaromatics such as imidazole and methylimidazole. The computed interaction energies are found to be in good agreement
Chih-Chin Tsou et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(47), 13358-13366 (2011-10-19)
S-nitrosation of the coordinated thiolate of dinitrosyl iron complexes (DNICs) to generate S-nitrosothiols (RSNOs) was demonstrated. Transformation of [{(NO)(2)Fe(μ-StBu)}(2)] (1-tBuS) into the {Fe(NO)(2)}(9) DNIC [(NO)(2)Fe(StBu)(MeIm)] (2-MeIm) occurs under addition of 20 equiv of 1-methylimidazole (MeIm) into a solution of 1-tBuS in
Fumitoshi Shibahara et al.
The Journal of organic chemistry, 77(19), 8815-8820 (2012-09-15)
Synthetic methods for triarylated azoles containing three different aryl groups via one-pot sequential multiple C-H bond arylations are described. The one-pot sequential diarylation of C5-monoarylated azoles was achieved by the simple sequential addition of two different aryl iodides with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service