Skip to Content
Merck
All Photos(2)

Documents

V900072

Sigma-Aldrich

Salicylic acid

Vetec, reagent grade, 98%

Synonym(s):

2-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2H
CAS Number:
Molecular Weight:
138.12
Beilstein:
774890
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

4.8 (vs air)

vapor pressure

1 mmHg ( 114 °C)

product line

Vetec

Assay

98%

form

powder, crystals or granules

bp

211 °C (lit.)

mp

158-161 °C (lit.)

density

1.44 g/cm3 at 20 °C

SMILES string

OC(=O)c1ccccc1O

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)

InChI key

YGSDEFSMJLZEOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E Benfeldt et al.
Acta dermato-venereologica, 79(5), 338-342 (1999-09-24)
Our aim was to simultaneously investigate 2 techniques for in vivo sampling of peripheral compartment pharmacokinetics after systemic administration of acetylsalicylic acid. Ten volunteers were given 2 g acetylsalicylic acid orally. Blood samples and dialysates from 4 microdialysis probes inserted
S Zaugg et al.
Journal of chromatography. B, Biomedical sciences and applications, 752(1), 17-31 (2001-03-20)
Acetylsalicylic acid (Aspirin) is rapidly metabolized to salicylic acid (salicylate) and other compounds, including gentisic acid and salicyluric acid. Monitoring of salicylate and its metabolites is of toxicological, pharmacological and biomedical interest. Three capillary electrophoresis (CE) methods featuring alkaline aqueous
Tsung-Meng Wu et al.
Plant molecular biology, 83(4-5), 379-390 (2013-06-21)
Glutathione reductases (GRs) are important components of the antioxidant machinery that plants use to respond against abiotic stresses. In rice, one cytosolic and two chloroplastic GR isoforms have been identified. In this work, we describe the cloning and characterization of
Dorothea Ellinger et al.
Plant physiology, 161(3), 1433-1444 (2013-01-22)
A common response by plants to fungal attack is deposition of callose, a (1,3)-β-glucan polymer, in the form of cell wall thickenings called papillae, at site of wall penetration. While it has been generally believed that the papillae provide a
Yunlong Du et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(19), 7946-7951 (2013-04-25)
Removal of cargos from the cell surface via endocytosis is an efficient mechanism to regulate activities of plasma membrane (PM)-resident proteins, such as receptors or transporters. Salicylic acid (SA) is an important plant hormone that is traditionally associated with pathogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service