Skip to Content
Merck
All Photos(2)

Documents

V900065

Sigma-Aldrich

Barium sulfate

Vetec, reagent grade, 99%

Synonym(s):

Baryte

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaSO4
CAS Number:
Molecular Weight:
233.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

form

powder

pH

7

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

SMILES string

[Ba++].[O-]S([O-])(=O)=O

InChI

1S/Ba.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

TZCXTZWJZNENPQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Craig Ungaro et al.
Optics express, 23(19), A1149-A1156 (2015-09-26)
This paper presents results on a highly efficient experimental solar thermophotovoltaic (STPV) system using simulated solar energy. An overall power conversion efficiency of 6.2% was recorded under solar simulation. This was matched with a thermodynamic model, and the losses within
Zhiyue Lin et al.
American journal of physiology. Gastrointestinal and liver physiology, 307(2), G158-G163 (2014-05-24)
This study aimed to develop and validate a method to measure bolus flow time (BFT) through the esophagogastric junction (EGJ) using a high-resolution impedance-manometry (HRIM) sleeve. Ten healthy subjects were studied with concurrent HRIM and videofluoroscopy; another 15 controls were
Rosie McDonald et al.
Journal of clinical pathology, 66(4), 330-334 (2013-01-05)
Faecal occult blood test (FOBt) is commonly requested by clinicians in both primary and secondary care. The aim of this study was to examine the use and outcomes of clinician-requested FOB testing in a single Trust taking part in the
Tafadzwa P Makarawo et al.
The American surgeon, 79(7), 728-733 (2013-07-03)
Water as an intraluminal negative contrast medium produces improved image quality with reduced artefact. However, rapid absorption of oral water in the bowel relative to speed and timing of image capturing has limited its clinical application. These findings predate advances
Koji Murono et al.
Diseases of the colon and rectum, 57(6), 715-724 (2014-05-09)
Preoperative chemoradiotherapy has been widely used for the prevention of local recurrence of locally advanced rectal cancer, and the effect of chemoradiotherapy is known to be associated with overall survival. We aimed to evaluate the association of the pathologic response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service