Skip to Content
Merck
All Photos(1)

Key Documents

V900039

Sigma-Aldrich

Iron(III) nitrate nonahydrate

Vetec, reagent grade, 98%

Synonym(s):

Ferric nitrate nonahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
404.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
reagent grade
form:
crystalline powder

grade

reagent grade

product line

Vetec

Assay

98%

form

crystalline powder

reaction suitability

reagent type: catalyst
core: iron

pH

1.5 (20 °C)

mp

47 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Fe.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SZQUEWJRBJDHSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guang-Li Wang et al.
Analytica chimica acta, 747, 92-98 (2012-09-19)
An innovative and versatile functional colorimetric sensor for melamine (MA) and H(2)O(2) was developed with simplicity, excellent selectivity and ultrasensitivity. The detection mechanism was based on the "oxidative etching-aggregation" of silver nanoparticles (AgNPs) by the cooperation effect of MA and
Sangchul Hwang et al.
Chemosphere, 78(5), 563-568 (2009-12-05)
Fenton-driven oxidation of methyl tert-butyl ether (MTBE) (0.11-0.16mM) in batch reactors containing ferric iron (5mM) and hydrogen peroxide (H(2)O(2)) (6mM) (pH=3) was performed to investigate MTBE transformation mechanisms. Independent variables included the forms of iron (Fe) (Fe(2)(SO(4))(3).9H(2)O and Fe(NO(3))(3).9H(2)O), H(2)O(2)
Milady Renata Apolinário da Silva et al.
Journal of hazardous materials, 146(3), 508-513 (2007-05-29)
The red Fe(2+)-phenanthroline complex is the basis of a classical spectrophotometric method for determination of iron. Due to the toxicity of this complexing agent, direct disposal of the wastewaters generated in analytical laboratories is not environmentally safe. This work evaluates
Jacob R Hascalovici et al.
Journal of neurochemistry, 108(1), 72-81 (2008-12-03)
Up-regulation of heme oxygenase-1 (HO-1) and altered cholesterol (CH) metabolism are characteristic of Alzheimer-diseased neural tissues. The liver X receptor (LXR) is a molecular sensor of CH homeostasis. In the current study, we determined the effects of HO-1 over-expression and
Mara Ghiazza et al.
Chemical research in toxicology, 24(1), 99-110 (2010-12-22)
The mechanism of enhancement/inhibition of quartz toxicity induced by iron is still unclear. Here the amount of iron on a fibrogenic quartz (Qz) was increased by wet impregnation (Fe(NO(3))(3) 0.67 and 6.7 wt %). X-ray diffraction (XRD), XRF diffuse reflectance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service