Skip to Content
Merck
All Photos(1)

Key Documents

V800436

Sigma-Aldrich

Trichloroethylene

LR, ≥99%

Synonym(s):

TCE, Trichloroethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CCl2
CAS Number:
Molecular Weight:
131.39
Beilstein:
1736782
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
grade:
LR
bp:
86.7 °C (lit.)
vapor pressure:
61 mmHg ( 20 °C)

grade

LR

vapor density

4.5 (vs air)

vapor pressure

61 mmHg ( 20 °C)

product line

Vetec

Assay

≥99%

form

liquid

autoignition temp.

770 °F

dilution

(for analytical testing)

refractive index

n20/D 1.476 (lit.)

bp

86.7 °C (lit.)

mp

−84.8 °C (lit.)

density

1.463 g/mL at 25 °C (lit.)

SMILES string

Cl\C=C(\Cl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H

InChI key

XSTXAVWGXDQKEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

closed cup - does not flash

Flash Point(C)

closed cup - does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda L Eiden et al.
Journal of medical entomology, 52(3), 429-436 (2015-09-04)
Permethrin is a commonly used acaricide for tick control on domestic animals and in residential environments, while fipronil use is restricted to on-animal treatment. Following widespread reports of permethrin and fipronil application failures to control indoor infestations of Rhipicephalus sanguineus
Glinda S Cooper et al.
Environmental health perspectives, 117(5), 696-702 (2009-05-30)
Our objective was to examine experimental and epidemiologic studies pertaining to immune-related, and specifically autoimmune-related, effects of trichloroethylene (TCE). We performed a literature search of PubMed and reviewed bibliographies in identified articles. We then systematically reviewed immune-related data, focusing on
Cheryl Siegel Scott et al.
Environmental health perspectives, 114(9), 1471-1478 (2006-09-13)
A large body of epidemiologic evidence exists for exploring causal associations between cancer and trichloroethylene (TCE) exposure. The U.S. Environmental Protection Agency 2001 draft TCE health risk assessment concluded that epidemiologic studies, on the whole, support associations between TCE exposure
Michihiro Kamijima et al.
International archives of occupational and environmental health, 80(5), 357-370 (2006-11-16)
Workers exposed to trichloroethylene (TCE) rarely show severe generalized skin disorders and accompanying hepatitis which resemble drug hypersensitivities. The disorders are completely different from solvent-induced irritating contact dermatitis, and their serious consequences have become one of the critical occupational health
P Cocco et al.
Occupational and environmental medicine, 70(11), 795-802 (2013-07-25)
We evaluated the association between occupational exposure to trichloroethylene (TCE) and risk of non-Hodgkin lymphoma (NHL) in a pooled analysis of four international case-control studies. Overall, the pooled study population included 3788 NHL cases and 4279 controls. Risk of NHL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service