Skip to Content
Merck
All Photos(1)

Key Documents

V800404

Sigma-Aldrich

Sodium thiosulfate

LR, ≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O3
CAS Number:
Molecular Weight:
158.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
crystalline powder

grade

LR

product line

Vetec

Assay

≥97%

form

crystalline powder

pH

6.0-8.5 (20 °C, 50 g/L)

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=S

InChI

1S/2Na.H2O3S2/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

AKHNMLFCWUSKQB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Van Haute et al.
International journal of food microbiology, 208, 102-113 (2015-06-13)
A methodology to i) assess the feasibility of water disinfection in fresh-cut leafy greens wash water and ii) to compare the disinfectant efficiency of water disinfectants was defined and applied for a combination of peracetic acid (PAA) and lactic acid
Michael C Reade et al.
Emergency medicine Australasia : EMA, 24(3), 225-238 (2012-06-08)
Cyanide poisoning is uncommon, but generates interest because of the presumed utility of an antidote immediately available in those areas with a high risk of cyanide exposure. As part of its regular review of guidelines, the Australian Resuscitation Council conducted
Melani Sooriyaarachchi et al.
Metallomics : integrated biometal science, 4(9), 960-967 (2012-07-31)
The anticancer drug cis-platin (CP) is widely used to treat patients, but it is also associated with significant side effects, including nephrotoxicity. Given that this metallodrug is intravenously (iv) administered, its biotransformations in the bloodstream are likely to be involved
Bita Arabshahi et al.
The Journal of pediatrics, 160(3), 520-522 (2012-01-17)
We report the successful use of abatacept and sodium thiosulfate in a patient with severe recalcitrant juvenile dermatomyositis complicated by ulcerative skin disease and progressive calcinosis. This combination therapy resulted in significant reductions in muscle and skin inflammation, decreased corticosteroid
Vikhyat S Bebarta et al.
Annals of emergency medicine, 59(6), 532-539 (2012-03-06)
We compare the efficacy of hydroxocobalamin to sodium thiosulfate to reverse the depressive effects on mean arterial pressure in a swine model of acute cyanide toxicity and gain a better understanding of the mechanism of action of the hydroxocobalamin in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service