Skip to Content
Merck
All Photos(1)

Key Documents

V800400

Sigma-Aldrich

Sodium sulfate

AR, anhydrous, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.5%
grade:
AR
anhydrous
form:
crystalline powder

grade

AR
anhydrous

product line

Vetec

Assay

≥99.5%

form

crystalline powder

pH

5.2-8.0 (20 °C, 50 g/L)

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

cation traces

Fe: ≤0.0005%
K: ≤0.002%
Mg: ≤0.001%
heavy metals (as Pb): ≤0.0005%

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ao Guo et al.
Scientific reports, 5, 14046-14046 (2015-09-16)
Regulatory T cells (Treg) play a critical role to control immune responses and to prevent autoimmunity, thus selective increase of Treg cells in vivo has broad therapeutic implications for autoimmune and inflammatory diseases. Licorice is a well-known herbal medicine used
Teppei Adachi et al.
Inflammatory bowel diseases, 21(1), 31-39 (2014-12-02)
Expression of heat shock protein A4 (HSPA4, also called Apg-2), a member of the HSP110 family, is induced by several forms of stress. The physiological and pathological functions of HSPA4 in the intestine remain to be elucidated. We assessed HSPA4
Arseto Y Bagastyo et al.
Water research, 47(1), 242-250 (2012-11-10)
Reverse osmosis concentrate from wastewater reclamation contains biorefractory trace organic contaminants that may pose environmental or health hazard. Due to its high conductivity, electrochemical oxidation of brine requires low voltage which is energetically favourable. However, the presence of chloride ions
Roosmarijn E Vandenbroucke et al.
Nature reviews. Drug discovery, 13(12), 904-927 (2014-11-08)
Matrix metalloproteinases (MMPs) are zinc-dependent endopeptidases that form a family of 24 members in mammals. Evidence of the pathological roles of MMPs in various diseases, combined with their druggability, has made them attractive therapeutic targets. Initial drug discovery efforts focused
Xiaorong Zhu et al.
The Journal of clinical investigation, 125(3), 1098-1110 (2015-02-03)
The intracellular protein HMGB1 is released from cells and acts as a damage-associated molecular pattern molecule during many diseases, including inflammatory bowel disease (IBD); however, the intracellular function of HMGB1 during inflammation is poorly understood. Here, we demonstrated that cytosolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service