Skip to Content
Merck
All Photos(2)

Key Documents

V800346

Sigma-Aldrich

p-Toluenesulfonic acid monohydrate

AR, ≥99%

Synonym(s):

4-Methylbenzenesulfonic acid monohydrate, 4-Toluenesulfonic acid monohydrate, p-Toluenesulfonic acid, PTSA monohydrate, TsOH monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3H · H2O
CAS Number:
Molecular Weight:
190.22
Beilstein:
3568023
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Pricing and availability is not currently available.

grade

AR

vapor density

5.9 (vs air)

product line

Vetec

Assay

≥99%

mp

103-106 °C (lit.)

functional group

tosylate

SMILES string

[H]O[H].Cc1ccc(cc1)S(O)(=O)=O

InChI

1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2

InChI key

KJIFKLIQANRMOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koneni V Sashidhara et al.
Bioorganic & medicinal chemistry letters, 22(12), 3926-3930 (2012-05-23)
First synthesis of novel coumarin-trioxane hybrids is reported. The synthesis was achieved via condensation of β-hydroxyhydroperoxides with coumarinic-aldehydes in presence of p-toluenesulfonic acid in good yields and the novel hybrids were evaluated for their antimalarial activity both in vitro and
Zhiqiang Ji et al.
Inorganic chemistry, 49(4), 1337-1346 (2010-01-23)
Two back-to-back terpyridine ligands using fluorenyl as bridging group (1-L and 2-L) and their corresponding dinuclear platinum(II) complexes (1 and 2) were synthesized and characterized. Their electronic absorption, photoluminescence, and the triplet transient difference absorption were systematically investigated. Both ligands
Aditya Kulkarni et al.
Organic letters, 13(19), 5124-5127 (2011-09-10)
An environmentally benign procedure for the hydrogenation of unprotected indoles is described. The hydrogenation reaction is catalyzed by Pt/C and activated by p-toluenesulfonic acid in water as a solvent. The efficacy of the method is illustrated by the hydrogenation of
Daniel C Yee et al.
The FEBS journal, 280(22), 5780-5800 (2013-08-29)
Visual rhodopsins are recognized members of the large and diverse family of G protein-coupled receptors (GPCRs), but their evolutionary origin and relationships to other proteins are not known. In a previous paper [Shlykov MA, Zheng WH, Chen JS & Saier
G Shanmugam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 95, 177-183 (2012-05-29)
Bulk crystals of newly identified organic nonlinear optical material 4-methylanilinium p-toluenesulfonate (PMPT) were grown by slow evaporation solution growth method using ethanol as a solvent. It crystallizes in monoclinic system with a noncentrosymmetric space group P2(1). The formation of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service