Skip to Content
Merck
All Photos(1)

Documents

V800341

Sigma-Aldrich

Potassium thiocyanate

LR, ≥97%

Synonym(s):

Potassium rhodanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KSCN
CAS Number:
Molecular Weight:
97.18
Beilstein:
3594799
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

LR

product line

Vetec

Assay

≥97%

form

crystalline powder

pH

5.3-8.7 (25 °C, 97.2 g/L)

mp

173 °C (lit.)

SMILES string

[K]SC#N

InChI

1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1

InChI key

ZNNZYHKDIALBAK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongtao Bian et al.
The journal of physical chemistry. B, 116(49), 14426-14432 (2012-11-28)
Microscopic structures and dynamics of aqueous salt solutions were investigated with the ultrafast vibrational energy exchange method and anisotropy measurements. In KSCN aqueous solutions of various concentrations, the rotational time constants of SCN(-) anions are proportional to the viscosities of
Altamir Benedito de Sousa et al.
Reproductive toxicology (Elmsford, N.Y.), 23(4), 568-577 (2007-03-06)
A study was performed at term and at weaning to verify the toxic effects of the prenatal exposure to potassium cyanide (KCN) and potassium thiocyanate (KSCN) in pregnant Wistar rats. Females received daily in drinking water the doses: 1, 3
Meiyun Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 75(3), 1013-1017 (2010-01-12)
A highly sensitive visible spectrophotometric method has been developed to determine ethamsylate in this paper, which is based on using Cu(II) as spectroscopic probe reagent. The study indicates that in the presence of SCN(-) and KNO(3), Cu(II) is reduced to
Yang-Hui Luo et al.
Dalton transactions (Cambridge, England : 2003), 43(44), 16937-16942 (2014-10-11)
A new iron(II) complex based on the 4,4'-dimethyl-2,2'-bipyridine ligand [Fe(4,4'-dmbpy)3(ClO4)(SCN)·3H2O (1·3H2O)] has been prepared and characterized. Structural studies and Hirshfeld surface analysis for complex 1·3H2O at three different temperatures (300, 240 and 130 K) are described. The UV-vis absorption spectrum
A Botti et al.
The journal of physical chemistry. B, 113(29), 10014-10021 (2009-07-03)
The results of a neutron diffraction experiment performed on aqueous solutions of KSCN are analyzed and discussed in comparison with similar data for KCl and KOH solutions. The effect of the different ions on the structure of water and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service