Skip to Content
Merck
All Photos(1)

Key Documents

V800298

Sigma-Aldrich

Petroleum ether

LR, bp 60-80 °C

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12191502
grade:
LR
bp:
30-60 °C (lit.)
60-80 °C
vapor pressure:
25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

grade

LR

vapor density

2.5 (vs air)

vapor pressure

25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

product line

Vetec

form

liquid

autoignition temp.

475 °F

expl. lim.

8 %

dilution

(for analytical testing)

refractive index

n20/D 1.363 (lit.)

bp

30-60 °C (lit.)
60-80 °C

density

0.64 g/mL at 25 °C (lit.)

SMILES string

[Mg+]c1ccc(cc1)C.[Br-]

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-56.2 °F - closed cup

Flash Point(C)

-49 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Dauger et al.
Archives de pediatrie : organe officiel de la Societe francaise de pediatrie, 10(3), 221-223 (2003-06-28)
The Authors report a case of acute White-Spirit poisoning with pulmonary hypertension associated to respiratory distress syndrome. A 14-month-old infant drank an unknown quantity of White-Spirit while his parents were painting. After he spontaneously vomited, he presented a seizure at
Lena Ernstgård et al.
Journal of applied toxicology : JAT, 29(3), 255-262 (2008-12-17)
A move from 'standard' white spirit (stdWS, 15-20% aromatics) to low-aromatic or dearomatized white spirit (deWS) has been seen, as the latter are considered to carry a smaller risk of health effects. However, data on health risks of deWS on
Gunnar Damgård Nielsen et al.
Basic & clinical pharmacology & toxicology, 98(2), 115-123 (2006-02-01)
Numerous studies have suggested that long-term occupational exposure to white spirit may cause chronic toxic encephalopathy (WHO 1996). This review summarizes the chronic nervous system effects of white spirit in animal studies during a 30-year period. First, routine histopathology was
H R Lam et al.
Toxicology letters, 80(1-3), 39-48 (1995-10-01)
The effects of 3 weeks' or 6 months' inhalation exposure of rats to aromatic white spirit 6 h/day, 5 days/week at 0, 400, or 800 ppm were studied. Synaptosomal neurochemistry was investigated as index of the in situ conditions in
Catherine F Poh et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(22), 6716-6722 (2006-11-24)
Genetically altered cells could become widespread across the epithelium of patients with oral cancer, often in clinically and histologically normal tissue, and contribute to recurrent disease. Molecular approaches have begun to yield information on cancer/risk fields; tissue optics could further

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service