Skip to Content
Merck
All Photos(1)

Key Documents

V800268

Sigma-Aldrich

N,N-Dimethylacetamide

AR, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737614
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
grade:
AR
bp:
164.5-166 °C (lit.)
vapor pressure:
2 mmHg ( 25 °C)
4 mmHg ( 38 °C)
Pricing and availability is not currently available.

grade

AR

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

product line

Vetec

Assay

≥99.5%

form

liquid

autoignition temp.

914 °F

expl. lim.

1.8 %, 100 °F
11.5 %, 160 °F

dilution

(for analytical testing)

refractive index

n20/D 1.437 (lit.)

pH

4 (20 °C, 200 g/L)

bp

164.5-166 °C (lit.)

mp

−20 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(C)=O

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inês Farinha et al.
Carbohydrate polymers, 130, 455-464 (2015-06-17)
Purified chitin-glucan complex (CGCpure) was extracted from Komagataella pastoris biomass using a hot alkaline treatment, followed by neutralization and repeated washing with deionized water. The co-polymer thus obtained had a β-glucan:chitin molar ratio of 75:25 and low protein and inorganic
Tony Reid et al.
The Lancet. Oncology, 16(9), 1133-1142 (2015-08-25)
Epigenetic alterations have been strongly associated with tumour formation and resistance to chemotherapeutic drugs, and epigenetic modifications are an attractive target in cancer research. RRx-001 is activated by hypoxia and induces the generation of reactive oxygen and nitrogen species that
S Sood et al.
Animal reproduction science, 129(1-2), 89-95 (2011-11-15)
Three experiments conducted to improve the survival of emu sperm during cryopreservation aimed to: (1) minimize chilling injury during the cooling phase; (2) determine the osmotic effects of dimethylacetamide (DMA), sucrose and trehalose; and (3) investigate the timing and nature
S Sood et al.
Theriogenology, 78(7), 1557-1569 (2012-09-18)
Three experiments were conducted to optimize the protocol for cryopreservation of emu sperm. Ejaculates were collected from trained male emus then diluted 1:1 and pooled before allocation to treatments and measured for sperm viability, motility, egg membrane penetration ability, membrane
Fen Ran et al.
Acta biomaterialia, 7(9), 3370-3381 (2011-06-11)
An amphiphilic triblock co-polymer of poly(vinyl pyrrolidone)-b-poly(methyl methacrylate)-b-poly(vinyl pyrrolidone) (PVP-b-PMMA-b-PVP) was synthesized via reversible addition-fragmentation chain transfer (RAFT) polymerization. The block co-polymer can be directly blended with polyethersulfone (PES) using dimethylacetamide (DMAC) as the solvent to prepare flat sheet and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service