Skip to Content
Merck
All Photos(1)

Key Documents

V800227

Supelco

2,2,4-Trimethylpentane

suitable for HPLC, ≥99.8%

Synonym(s):

Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
Assay:
≥99.8%
technique(s):
HPLC: suitable
bp:
98-99 °C (lit.)
vapor pressure:
41 mmHg ( 21 °C)
Pricing and availability is not currently available.

vapor density

3.9 (vs air)

vapor pressure

41 mmHg ( 21 °C)

product line

Vetec

Assay

≥99.8%

form

liquid

autoignition temp.

745 °F

expl. lim.

6 %

technique(s)

HPLC: suitable

refractive index

n20/D 1.391 (lit.)

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

density

0.692 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Swarna Bansal et al.
Journal of hazardous materials, 190(1-3), 45-50 (2011-04-05)
Separation of secondary emulsions of dispersed droplet size less than 10 μm, by means of fibrous medium is a very complex but important process. The study investigates the influence of thin fibrous filter properties, i.e. surface energy, pore size and
Yufeng Zhang et al.
Journal of chromatography. A, 1252, 67-73 (2012-07-10)
In this study, a polyethylene Pasteur pipette-based ultrasound-assisted emulsification microextraction (USAEME) applying low-density organic solvent was successfully developed for the extraction of trace levels of organochlorine pesticides (OCPs) in water samples and followed by gas chromatography-mass spectrometry (GC-MS) analysis. In
Kyung Min Park et al.
Food chemistry, 138(2-3), 733-738 (2013-02-16)
In this study, we investigated the possibility of reversed micelles, widely used as an enzyme reactor for lipases, for the determination of lipoxygenase activity. Although it is rapid and simple, reversed micelles have some limitations, such as interference by UV-absorbing
Aram Kang et al.
Molecular bioSystems, 8(4), 1350-1358 (2012-02-14)
Microbial tolerance to hydrocarbons has been studied in an effort to improve the productivity of biochemical processes and to enhance the efficiency of hydrocarbon bioremediation. Despite these studies, few attempts have been made to design rational strategies to improve microbial
Koichi Sawada et al.
Journal of bioscience and bioengineering, 114(2), 204-208 (2012-05-19)
Electrospun polyvinyl alcohol (PVA) nanofibrous non-woven fabrics have been widely used for cell and enzyme immobilization. Enhancement of the productivity of the material will further enlarge the versatility of them. In this study, a mixture of water and N,N-dimethylformamide (DMF)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service