Skip to Content
Merck
All Photos(1)

Key Documents

V800199

Sigma-Aldrich

Hexamethylenetetramine

LR, ≥99%

Synonym(s):

1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, Hexamine, Methenamine, Urotropine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N4
CAS Number:
Molecular Weight:
140.19
Beilstein:
2018
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

LR

vapor pressure

<0.01 mmHg ( 20 °C)

product line

Vetec

Assay

≥99%

form

crystalline powder

autoignition temp.

770 °F

pH

8.5-9.5 (10% in water)

mp

280 °C (subl.) (lit.)

SMILES string

C1N2CN3CN1CN(C2)C3

InChI

1S/C6H12N4/c1-7-2-9-4-8(1)5-10(3-7)6-9/h1-6H2

InChI key

VKYKSIONXSXAKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Skin Sens. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

482.0 °F - closed cup

Flash Point(C)

250 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ann Pontén et al.
Contact dermatitis, 68(1), 50-53 (2012-10-06)
The currently used patch test concentration for formaldehyde is 1.0% (wt/vol) in water. However, clinical experience and previous studies suggest that 1.0% might be insufficient for detecting an optimized number of clinically relevant cases of contact allergy to formaldehyde. To
Jianguo Hu et al.
Talanta, 93, 345-349 (2012-04-10)
A green, one-step method for synthesis of graphene-Au nanoparticles (graphene-AuNPs) was introduced in this article, using an environmentally benign hexamethylenetetramine (HMTA) as reducing and stabilizing agent. HMTA slowly was hydrolyzed to generate aldehyde ammonia to reduce graphene oxides (GO) and
Hong Yu et al.
Nanoscale, 6(18), 10556-10561 (2014-08-15)
Ternary metal oxides have been receiving wide attention in electrochemical energy storage due to their rich redox reactions and tuneable conductivity. We present a simple solution-based method to prepare a 3D interconnected porous network of ternary metal oxide (CoMoO₄ and
Vincent Consonni et al.
ACS nano, 8(5), 4761-4770 (2014-04-12)
Controlling the polarity of ZnO nanowires in addition to the uniformity of their structural morphology in terms of position, vertical alignment, length, diameter, and period is still a technological and fundamental challenge for real-world device integration. In order to tackle
Andreas Lemmerer et al.
Chemical communications (Cambridge, England), 48(13), 1883-1885 (2011-12-03)
The tetrahedral arrangement of hydrogen bonding donor and acceptor groups is used to rationalise the design of a diamondoid network; however, a single proton transfer renders the four sites inequivalent, and results in two polymorphs of the title molecular salt

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service