V800041
Ammonium chloride
AR, ≥99.8%
Synonym(s):
Salmiac
About This Item
Recommended Products
grade
AR
vapor density
1.9 (vs air)
vapor pressure
1 mmHg ( 160.4 °C)
product line
Vetec™
Assay
≥99.8%
form
crystalline powder
pH
4.6-6 (5% in water)
mp
340 °C (subl.) (lit.)
anion traces
sulfate (SO42-): ≤0.002%
cation traces
Fe: ≤0.0002%
heavy metals (as Pb): ≤0.0002%
SMILES string
N.Cl
InChI
1S/ClH.H3N/h1H;1H3
InChI key
NLXLAEXVIDQMFP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service