Skip to Content
Merck
All Photos(1)

Key Documents

V800013

Sigma-Aldrich

2-Ethoxyethanol

LR, ≥99%

Synonym(s):

Ethyl glycol, Ethylene glycol monoethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1098271
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
LR
bp:
135 °C (lit.)
vapor pressure:
3.8 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

LR

vapor density

3.1 (vs air)

vapor pressure

3.8 mmHg ( 20 °C)

product line

Vetec

Assay

≥99%

form

liquid

autoignition temp.

460 °F

expl. lim.

14 %

dilution

(for analytical testing)

refractive index

n20/D 1.407 (lit.)

bp

135 °C (lit.)

mp

−90 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

SMILES string

CCOCCO

InChI

1S/C4H10O2/c1-2-6-4-3-5/h5H,2-4H2,1H3

InChI key

ZNQVEEAIQZEUHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui-Sheng Wang et al.
Industrial health, 45(4), 574-578 (2007-09-20)
Ethylene glycol monoethyl ether (EGEE) can cause damage to testes and sperm, and its metabolites are believed to play an important role in its toxicity. Aldehyde dehydrogenase 2 (ALDH2) is involved in the metabolism of this chemical. To investigate whether
Hong Soon Choi et al.
Journal of occupational health, 46(4), 260-265 (2004-08-17)
Alkyl cellosolves include ethylene glycol monomethylether, ethylene glycol monoethylether, ethylene glycol monobuthylether. And their urine metabolites are methoxyacetic acid, ethoxyacetic acid and butoxyacetic acid. The current analytical method for urinary alkoxyacetic acid is liquid-liquid phase extraction. But the liquid-liquid phase
Matthew J Traynor et al.
Toxicology and applied pharmacology, 218(2), 128-134 (2006-12-19)
Glycol ethers are solvents widely used alone and as mixtures in industrial and household products. Some glycol ethers have been shown to have a range of toxic effects in humans following absorption and metabolism to their aldehyde and acid metabolites.
David J Lockley et al.
Toxicology and applied pharmacology, 180(2), 74-82 (2002-04-24)
Percutaneous absorption and cutaneous metabolism of 2-ethoxyethanol were assessed in vivo and with an in vitro flow-through diffusion system. Topical application of undiluted (14)C-ethoxyethanol to occluded rat skin in vivo resulted in 25% of the dose being absorbed after 24
I A Adedara et al.
Andrologia, 45(2), 111-119 (2012-06-20)
Endocrine disrupting chemicals cause reproductive dysfunction by interacting with intricate regulation and cellular processes involve in spermatogenesis. This study investigated the probable mechanism of action of ethylene glycol monoethyl ether (EGEE) as an antiandrogenic compound as well as the effects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service