Skip to Content
Merck
All Photos(1)

Key Documents

V000217

Sigma-Aldrich

Mercury(II) sulfate

ACS reagent, 98%

Synonym(s):

Mercuric sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgSO4
CAS Number:
Molecular Weight:
296.65
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

ACS reagent

Assay

98%

SMILES string

[Hg++].[O-]S([O-])(=O)=O

InChI

1S/Hg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

DOBUSJIVSSJEDA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shovana Pradhan et al.
Chemosphere, 136, 198-203 (2015-05-24)
Reverse osmosis (RO) concentrate (ROC) streams generated from RO-based municipal wastewater reclamation processes pose potential health and environmental risks on their disposal to confined water bodies such as bays. A UV/H2O2 advanced oxidation process followed by a biological activated carbon
K Kawai et al.
Nuclear medicine and biology, 22(1), 37-43 (1995-01-01)
Fluorodemercuration has the greatest utility for the preparation of 6-[18F]DOPA, but requires separation from unreacted mercury precursor and other mercury-containing compounds. One approach is the development of a polymer-bound mercury precursor. In this study, polymer-bound 6-thiolatomercury and 6-mercuric sulfonate DOPA
A M Abdelhamid
Archiv fur Tierernahrung, 38(3), 207-214 (1988-03-01)
This work was designed to study effects of HgSO4 on rabbits performance when added in the mash diet for 7 weeks with concentrations of 0, 150, and 300 ppm as Hg (6 animals/group). The Hg application caused mortality associated with
Necati Kayaalp et al.
Environmental science and pollution research international, 17(9), 1547-1552 (2010-05-25)
Chemical oxygen demand (COD) is used as a discharge standard parameter in wastewater treatment plant design, environmental modelling and many other applications. Chloride interference is an important problem of COD measurement for wastewaters containing low organic matter and high chloride
A P de Souza et al.
Dental materials : official publication of the Academy of Dental Materials, 16(2), 103-108 (2001-02-24)
The interaction between metal ions and the oral environment is a major subject matter in dental research. Matrix metalloproteinases (MMPs) have been implicated in several pathologic oral processes such as periodontal tissue destruction, root caries, tumour invasion and temporomandibular joint

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service