Skip to Content
Merck

1A00880

USP

11β,16a,17,21-Tetrahydroxypregna-1,4-dien-3,20-dione

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

11β,16α,17,21-Tetrahydroxypregna-1,4-dien-3,20-dione, (8S,9S,10R,11S,13S,14S,16R,17S)-11,16,17-trihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-3-one, 16α-Hydroxyprednisolone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H28O6
CAS Number:
Molecular Weight:
376.44
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical analytical impurity (PAI)

agency

USP

API family

prednisolone

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

O[C@]1([C@@]2([C@H]([C@H]3[C@@H]([C@@]4(C(=CC(=O)C=C4)CC3)C)[C@H](C2)O)C[C@H]1O)C)C(=O)CO

InChI key

SEKYBDYVXDAYPY-ILNISADRSA-N

General description

11β,16α,17,21-Tetrahydroxypregna-1,4-dien-3,20-dione is a USP Pharmaceutical Analytical Impurity (PAI).

USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.

Associated Drug Substance: Budesonide

Therapeutic Area: Steroids

For more information about this PAI, visit here.

Application

11β,16α,17,21-Tetrahydroxypregna-1,4-dien-3,20-dione (USP PAI) is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development. PAI can be used in the following applications:

1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library