Skip to Content
Merck

Skip To

1711075

USP

Vardenafil Related Compound E

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(2-[2-Ethoxy-5-(piperazin-1-ylsulfonyl)phenyl]-5-methyl-7-propylimidazo[5,1-f] [1,2,4]triazin-4(3H)-one)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 1.360,00

About This Item

Empirical Formula (Hill Notation):
C21H28N6O4S
CAS Number:
Molecular Weight:
460.55
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904

€ 1.360,00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

vardenafil

packaging

pkg of 25 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)
pharmaceutical
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N4CCNCC4)c1cc(c(cc1)OCC)c2[nH][n]3c([c](n2)=O)c(nc3CCC)C

InChI

1S/C21H28N6O4S/c1-4-6-18-23-14(3)19-21(28)24-20(25-27(18)19)16-13-15(7-8-17(16)31-5-2)32(29,30)26-11-9-22-10-12-26/h7-8,13,22H,4-6,9-12H2,1-3H3,(H,24,25,28)

InChI key

OKUKNGDVADFTHE-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Vardenafil Related Compound E USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Vardenafil Orally Disintegrating Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
17110641711029
description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

packaging

pkg of 25 mg

packaging

pkg of 25 mg

packaging

-

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP


Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2 - STOT RE 2

Target Organs

Cardio-vascular system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters