Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
15 mg | Check Cart for Availability | € 1.120,00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
valacyclovir
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
pharmaceutical
pharmaceutical
format
neat
storage temp.
2-8°C
General description
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| format neat | format neat | format neat | format neat |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| API family valacyclovir | API family valacyclovir | API family abacavir | API family abacavir |
| agency USP | agency USP | agency USP | agency USP |
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

