Skip to Content
Merck

Skip To

1705367

USP

Ulipristal acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(11β)-17-(Acetyloxy)-11-[4-(dimethylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione, CDB-2914

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Check Cart for Availability
€ 811,00

About This Item

Empirical Formula (Hill Notation):
C30H37NO4
CAS Number:
Molecular Weight:
475.62
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

€ 811,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

USP

API family

ulipristal

packaging

pkg of 200 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=C(C)[C@@]1(OC(C)=O)CC[C@@]2([H])[C@]3([H])CCC4=CC(CCC4=C3[C@@H](C5=CC=C(N(C)C)C=C5)C[C@@]21C)=O

InChI

1S/C30H37NO4/c1-18(32)30(35-19(2)33)15-14-27-25-12-8-21-16-23(34)11-13-24(21)28(25)26(17-29(27,30)3)20-6-9-22(10-7-20)31(4)5/h6-7,9-10,16,25-27H,8,11-15,17H2,1-5H3/t25-,26+,27-,29-,30-/m0/s1

InChI key

OOLLAFOLCSJHRE-ZHAKMVSLSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ulipristal acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Ulipristal acetate is a selective progesterone receptor modulator (SPRM) with tissue-selective partial antagonist activity. It has clinical use both as an emergency contraceptive and as a treatment for uterine fibroids. Ulipristal acetate acts as a partial antagonist on the hypothalamic–pituitary–ovarian axis to inhibit or delay ovulation without affecting human embryo implantation. As a progesterone antagonist, Ulipristal acetate selectively suppresses neo-vascularization, cell proliferation, and survival in uterine fibroid cells, but not in normal myometrial cells.
Ulipristal acetate is a selective progesterone receptor modulator (SPRM); contraceptive and treatment for fibroid tumors.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML146917053781166535
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

manufacturer/tradename

USP

format

neat

format

-

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

packaging

pkg of 200 mg

packaging

-

packaging

pkg of 25 mg

packaging

pkg of 20 mg

API family

ulipristal

API family

-

API family

ulipristal

API family

norethindrone

grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard


Still not finding the right product?

Explore all of our products under Ulipristal acetate


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters