Skip to Content
Merck
All Photos(1)

Key Documents

1704003

USP

Tyloxapol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-(1,1,3,3-Tetramethylbutyl)phenol polymer with formaldehyde and oxirane

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 200 MG
€404.00

€404.00


Estimated to ship on09 April 2025


Request a Bulk Order

Select a Size

Change View
3 X 200 MG
€404.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€404.00


Estimated to ship on09 April 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

tyloxapol

manufacturer/tradename

USP

CMC

0.018 mM

transition temp

cloud point 94.3 °C

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H]C([H])=O.OCCO.CC(C)(C)CC(C)(C)c1ccc(O)cc1

InChI

1S/C14H22O.C2H6O2.CH2O/c1-13(2,3)10-14(4,5)11-6-8-12(15)9-7-11;3-1-2-4;1-2/h6-9,15H,10H2,1-5H3;3-4H,1-2H2;1H2

InChI key

GWJOFBXSBDVUMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A nonionic liquid polymer of the alkyl aryl polyether alcohol type. Used as a surfactant.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tyloxapol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service