Skip to Content
Merck

Skip To

1699956

USP

Trypsin Bovine

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
300 mg
Check Cart for Availability
€ 829,00

About This Item

Empirical Formula (Hill Notation):
C1012H1585N279O324S14
Molecular Weight:
23293.00
NACRES:
NA.78
UNSPSC Code:
41116107

€ 829,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

form

solid

mol wt

23,293 Da (for β-Trypsin)

packaging

pkg of 30 mg

manufacturer/tradename

USP

application(s)

USP Biologics

format

neat

storage temp.

-10 to -25°C

General description

Bovine-derived trypsin is a serine protease that specifically cleaves peptide bonds at the carboxyl side of lysine and arginine residues. The USP Trypsin Bovine standard ensures enzymatic activity and purity, making it suitable for tissue dissociation, proteomics workflows, and cell culture applications.

The USP biologics proteins category provides critical standards and reference materials necessary for the development and manufacturing of therapeutic proteins. These proteins, which can be naturally derived or genetically engineered, are essential in treating various diseases and conditions, ensuring that they meet quality, safety, and efficacy standards.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Trypsin Bovine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Chymotrypsin
  • Chymotrypsin for Ophthalmic Solution
  • Trypsin

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
This product is part of the USP Biologics program.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
143702013424011165204
application(s)

USP Biologics

application(s)

pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

form

solid

form

-

form

liquid

form

-

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

-65 to -96°C

storage temp.

2-8°C

packaging

pkg of 30 mg

packaging

pkg of 50 mg

packaging

pkg of 1 mL

packaging

pkg of 100 mg

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters