Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1.82 mg | Check Cart for Availability | € 498,00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
triptorelin
form
solid
manufacturer/tradename
USP
application(s)
USP Biologics
pharmaceutical
storage temp.
−20°C
General description
Peptide family: Triptorelin
A GnRH analog important for treating hormone-sensitive cancers.
The USP biologics peptides category encompasses a diverse range of therapeutic peptides that are essential in managing various medical conditions. These peptides, typically consisting of amino acid sequences of 40 residues or less, are critical for the development of high-quality medicines. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of peptide therapeutics throughout their lifecycle.
The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| application(s) USP Biologics | application(s) USP Biologics | application(s) USP Biologics | application(s) USP Biologics |
| form solid | form solid | form powder | form solid |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
| API family triptorelin | API family triptorelin | API family goserelin | API family cosyntropin |
signalword
Danger
hcodes
pcodes
Hazard Classifications
Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


