Skip to Content
Merck
All Photos(1)

Documents

1683537

USP

Triethyl Aconitate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(Triethyl (E)-prop-1-ene-1,2,3-tricarboxylate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O6
CAS Number:
Molecular Weight:
258.27
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

USP

manufacturer/tradename

USP 1683537

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

InChI

1S/C12H18O6/c1-4-16-10(13)7-9(12(15)18-6-3)8-11(14)17-5-2/h7H,4-6,8H2,1-3H3/b9-7+

InChI key

IDDWGDKSBYYEPL-VQHVLOKHSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Triethyl Aconitate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Triethyl Citrate
  • Acetyltriethyl Citrate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

USP issued SDS can be found here.

Other Notes

Sales restrictions may apply.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service