Skip to Content
Merck

Skip To

1643306

USP

Tamoxifen citrate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Tamoxifen citrate salt, (Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Check Cart for Availability
€ 407,00

About This Item

Empirical Formula (Hill Notation):
C26H29NO · C6H8O7
CAS Number:
Molecular Weight:
563.64
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

€ 407,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

tamoxifen

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(c1ccccc1)=C(/c2ccccc2)c3ccc(OCCN(C)C)cc3

InChI

1S/C26H29NO.C6H8O7/c1-4-25(21-11-7-5-8-12-21)26(22-13-9-6-10-14-22)23-15-17-24(18-16-23)28-20-19-27(2)3;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-18H,4,19-20H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b26-25-;

InChI key

FQZYTYWMLGAPFJ-OQKDUQJOSA-N

Gene Information

human ... ESR1(2099)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tamoxifen citrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Tamoxifen Citrate Tablets

Biochem/physiol Actions

Estrogen antagonist in mammary gland. Blocks estradiol-stimulated VEGF production in breast tumor cells. Protein kinase C inhibitor.
Protein kinase C inhibitor. Induces apoptosis in human malignant glioma cell lines. Tamoxifen and its metabolite 4-hydroxytamoxifen are selective estrogen response modifiers (SERMs) that act as estrogen antagonists in mammary gland. Blocks estradiol-stimulated VEGF production in breast tumor cells.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T0014000T0015000BP1225
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

EDQM

manufacturer/tradename

BP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

Gene Information

human ... ESR1(2099)

Gene Information

human ... ESR1(2099)

Gene Information

human ... ESR1(2099)

Gene Information

human ... ESR1(2099)

API family

tamoxifen

API family

tamoxifen

API family

tamoxifen

API family

tamoxifen


Still not finding the right product?

Explore all of our products under Tamoxifen citrate


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters