Skip to Content
Merck
All Photos(1)

Key Documents

1642100

USP

Sulisobenzone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5-Benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid, HMBS, Sulisobenzone

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€423.00

€423.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
500 MG
€423.00

About This Item

Empirical Formula (Hill Notation):
C14H12O6S
CAS Number:
Molecular Weight:
308.31
Beilstein:
2889165
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€423.00


Estimated to ship on20 May 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

sulisobenzone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1cc(O)c(cc1S(O)(=O)=O)C(=O)c2ccccc2

InChI

1S/C14H12O6S/c1-20-12-8-11(15)10(7-13(12)21(17,18)19)14(16)9-5-3-2-4-6-9/h2-8,15H,1H3,(H,17,18,19)

InChI key

CXVGEDCSTKKODG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulisobenzone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

René Böttcher et al.
Oncotarget, 6(6), 4036-4050 (2015-02-18)
Current prostate cancer (PCa) biomarkers such as PSA are not optimal in distinguishing cancer from benign prostate diseases and predicting disease outcome. To discover additional biomarkers, we investigated PCa-specific expression of novel unannotated transcripts. Using the unique probe design of
Kamila Kyrova et al.
BMC veterinary research, 10, 244-244 (2014-10-02)
Following infection and initial multiplication in the gut lumen, Salmonella Typhimurium crosses the intestinal epithelial barrier and comes into contact with cells of the host immune system. Mononuclear phagocytes which comprise macrophages and dendritic cells (DC) are of key importance
Marjolein Bijlard et al.
Molecular and cellular biology, 35(4), 675-687 (2014-12-17)
Myelination of axons by oligodendrocytes is essential for saltatory nerve conduction. To form myelin membranes, a coordinated synthesis and subsequent polarized transport of myelin components are necessary. Here, we show that as part of the mechanism to establish membrane polarity
David C Schöndorf et al.
Nature communications, 5, 4028-4028 (2014-06-07)
Mutations in the acid β-glucocerebrosidase (GBA1) gene, responsible for the lysosomal storage disorder Gaucher's disease (GD), are the strongest genetic risk factor for Parkinson's disease (PD) known to date. Here we generate induced pluripotent stem cells from subjects with GD

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service