Skip to Content
Merck

Skip To

1612459

USP

Sevelamer carbonate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Poly(allylamine-co-N,N′-diallyl-1,3-diamino-2-hydroxypropane) carbonate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
[(C3H7N)a+b(C9H18N2O)c]n · x H2CO3
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

sevelamer

packaging

pkg of 100 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

ClCC1OC1.[N+H3]CC=C.[O-]C(=O)O

InChI

1S/C3H5ClO.C3H7N.CH2O3/c4-1-3-2-5-3;1-2-3-4;2-1(3)4/h3H,1-2H2;2H,1,3-4H2;(H2,2,3,4)

InChI key

PADGNZFOVSZIKZ-UHFFFAOYSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Sevelamer carbonate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
107578010123161A01540
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecules)

application(s)

pharmaceutical

format

neat

format

neat

format

neat

format

neat

API family

sevelamer

API family

bisoprolol

API family

methionine & derivatives

API family

-

packaging

pkg of 100 mg

packaging

-

packaging

pkg of 250 mg

packaging

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters