Skip to Content
Merck

1601747

USP

Residual Solvent Class 2 - Pyridine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Pyridine solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

SMILES string

n1ccccc1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Residual Solvent Class 2 - Pyridine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

188.6 °F

Flash Point (°C)

87.00 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

110-86-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library