Skip to Content
Merck

Skip To

1548076

USP

Polyvinyl chloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Poly(vinyl chloride), PVC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 407,00

About This Item

Linear Formula:
(CH2CHCl)n
CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

€ 407,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

density

1.4 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

ClC=C

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2

InChI key

BZHJMEDXRYGGRV-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Polyvinyl chloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8138881387346764
description

United States Pharmacopeia (USP) Reference Standard

description

low molecular weight

description

high molecular weight

description

average Mw ~233,000, average Mn ~99,000

grade

pharmaceutical primary standard

grade

-

grade

-

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

-

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

battery manufacturing

application(s)

battery manufacturing

density

1.4 g/mL at 25 °C (lit.)

density

1.4 g/mL at 25 °C (lit.)

density

1.4 g/mL at 25 °C (lit.)

density

1.4 g/mL at 25 °C (lit.)

agency

USP

agency

-

agency

-

agency

-


Still not finding the right product?

Explore all of our products under Polyvinyl chloride


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the nominal particle size of this product?

    1 answer
    1. The particle size of this material is not determined.

      Helpful?

Reviews

No rating value

Active Filters