Skip to Content
Merck

1542003

USP

Piperazine phosphate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Piperazinium dihydrogen phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C4H10N2 · H3PO4
CAS Number:
Molecular Weight:
184.13
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Piperazine phosphate, United States Pharmacopeia (USP) Reference Standard

InChI

1S/C4H10N2.H3O4P/c1-2-6-4-3-5-1;1-5(2,3)4/h5-6H,1-4H2;(H3,1,2,3,4)

InChI key

NQQWFVUVBGSGQN-UHFFFAOYSA-N

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service