Skip to Content
Merck
All Photos(1)

Key Documents

1504955

USP

1-Pentanol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

n-Amyl alcohol, Pentyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 1.2 ML
€343.00

€343.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
3 X 1.2 ML
€343.00

About This Item

Linear Formula:
CH3(CH2)4OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730975
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€343.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

pharmaceutical primary standard

vapor density

3 (vs air)

vapor pressure

1 mmHg ( 13.6 °C)
1.5 mmHg ( 20 °C)

form

liquid

expl. lim.

10 %, 100 °F

manufacturer/tradename

USP

refractive index

n20/D 1.409 (lit.)

bp

136-138 °C (lit.)

mp

−78 °C (lit.)

density

0.811 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCO

InChI

1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

AMQJEAYHLZJPGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

1-Pentanol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service