Skip to Content
Merck
All Photos(1)

Key Documents

1494807

USP

Pantolactone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-(−)-Pantolactone, (R)-(−)-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, (R)-(−)-β,β-Dimethyl-α-hydroxy-γ-butyrolactone, (R)-(−)-Pantolactone, Pantoic acid γ-lactone

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€1,140.00

€1,140.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
500 MG
€1,140.00

About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
Beilstein:
80957
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€1,140.00


Estimated to ship on24 May 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

pantolactone

manufacturer/tradename

USP

bp

120-122 °C/15 mmHg (lit.)

mp

91 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)COC(=O)[C@@H]1O

InChI

1S/C6H10O3/c1-6(2)3-9-5(8)4(6)7/h4,7H,3H2,1-2H3/t4-/m0/s1

InChI key

SERHXTVXHNVDKA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pantolactone USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Dexpanthenol Preparation
  • Calcium Pantothenate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service