Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
10 mg | Check Cart for Availability | € 1.440,00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
paclitaxel
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
mixture
storage temp.
2-8°C
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename EDQM |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| format mixture | format mixture | format mixture | format neat |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C |
| API family paclitaxel | API family docetaxel | API family valrubicin | API family pemetrexed |
signalword
Danger
hcodes
Hazard Classifications
Muta. 2 - Repr. 1B - STOT RE 1
target_organs
Central nervous system,Bone marrow,Cardio-vascular system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

